Please login to your online account to display your discounted pricing

N,N-Diformylacetamide 80%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC380450010

 View more versions of this product

Catalog No. AC380450010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥75%
Physical Form Liquid
Formula Weight 115.09
Quantity 1g
Color Clear colorless
CAS Min % 75.0
CAS Max % 100.0
Linear Formula CH3C(O)N(COH)2
MDL Number MFCD00015916
CAS 26944-31-0
Synonym Acetyldiformamide
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Chemical Name or Material N, N-Diformylacetamide
Assay Percent Range 80%
Molecular Formula C4H5NO3
Infrared Spectrum Authentic

Specifications

Chemical Name or Material N, N-Diformylacetamide
CAS 26944-31-0
CAS Min % 75.0
CAS Max % 100.0
Assay ≥75%
Assay Percent Range 80%
Molecular Formula C4H5NO3
Linear Formula CH3C(O)N(COH)2
Formula Weight 115.09
MDL Number MFCD00015916
Synonym Acetyldiformamide
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents