missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N-Diformylacetamide 80%, ACROS Organics™

Chemical Identifiers

CAS 26944-31-0
CAS Min % 75.0
CAS Max % 100.0
Assay Percent Range 80%
Molecular Formula C4H5NO3
Linear Formula CH3C(O)N(COH)2
Formula Weight 115.09
MDL Number MFCD00015916
Synonym Acetyldiformamide
Chemical Name or Material N, N-Diformylacetamide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC380450050 SDS Thermo Fisher Scientific
AC380450050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC380450010 SDS Thermo Fisher Scientific
AC380450010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

26944-31-0
100.0
C4H5NO3
115.09
Acetyldiformamide
≥75%
75.0
80%
CH3C(O)N(COH)2
MFCD00015916
N, N-Diformylacetamide

Specifications

Clear colorless
26944-31-0
100.0
Glass bottle
80%
CH3C(O)N(COH)2
MFCD00015916
N, N-Diformylacetamide
5g
75.0
Authentic
Liquid
C4H5NO3
115.09
Acetyldiformamide
≥75%

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.