missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Diethylsalicylamide, 97%

Manufacturer:  Alfa Aesar™ H5502506

 View more versions of this product

Catalog No. AAH5502506



Description

Description

Applications
N,N-Diethylsalicylamide is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

N,N-Diethylsalicylamide
5g
C11H15NO2
2-hydroxy-n,n-diethylbenzamide
CCN(CC)C(=O)C1=CC=CC=C1O
193.246
193.25
93°C to 97°C
19311-91-2
MFCD00010816
ZVYXEXAXXWINEH-UHFFFAOYSA-N
N,N-diethyl-2-hydroxybenzamide
88006
97%

Chemical Identifiers

19311-91-2
193.246
ZVYXEXAXXWINEH-UHFFFAOYSA-N
88006
CCN(CC)C(=O)C1=CC=CC=C1O
C11H15NO2
MFCD00010816
2-hydroxy-n,n-diethylbenzamide
N,N-diethyl-2-hydroxybenzamide
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : VN7320000

Documents