Learn More
N,N-Diethylsalicylamide, 97%, Thermo Scientific Chemicals
$59.42 - $196.01
Chemical Identifiers
CAS | 19311-91-2 |
---|---|
Molecular Formula | C11H15NO2 |
Molecular Weight (g/mol) | 193.246 |
MDL Number | MFCD00010816 |
InChI Key | ZVYXEXAXXWINEH-UHFFFAOYSA-N |
Synonym | n,n-diethylsalicylamide, diethylsalicylamide, salicyldiethylamide, salicylamide, n,n-diethyl, benzamide, n,n-diethyl-2-hydroxy, o-hydroxy-n,n-diethylbenzamide, unii-hg05mo88tl, hg05mo88tl, acmc-20ak91, 2-hydroxy-n,n-diethylbenzamide |
PubChem CID | 88006 |
IUPAC Name | N,N-diethyl-2-hydroxybenzamide |
SMILES | CCN(CC)C(=O)C1=CC=CC=C1O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5502503
|
Thermo Scientific Chemicals
H5502503 |
1 g |
N/A
|
|
|||||
AAH5502506
|
Thermo Scientific Chemicals
H5502506 |
5 g |
N/A
|
|
|||||
Description
N,N-Diethylsalicylamide is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN,N-Diethylsalicylamide is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
19311-91-2 | |
193.246 | |
ZVYXEXAXXWINEH-UHFFFAOYSA-N | |
88006 | |
CCN(CC)C(=O)C1=CC=CC=C1O |
C11H15NO2 | |
MFCD00010816 | |
n,n-diethylsalicylamide, diethylsalicylamide, salicyldiethylamide, salicylamide, n,n-diethyl, benzamide, n,n-diethyl-2-hydroxy, o-hydroxy-n,n-diethylbenzamide, unii-hg05mo88tl, hg05mo88tl, acmc-20ak91, 2-hydroxy-n,n-diethylbenzamide | |
N,N-diethyl-2-hydroxybenzamide |
Specifications
19311-91-2 | |
C11H15NO2 | |
1 g | |
ZVYXEXAXXWINEH-UHFFFAOYSA-N | |
N,N-diethyl-2-hydroxybenzamide | |
88006 | |
97% |
93°C to 97°C | |
MFCD00010816 | |
n,n-diethylsalicylamide, diethylsalicylamide, salicyldiethylamide, salicylamide, n,n-diethyl, benzamide, n,n-diethyl-2-hydroxy, o-hydroxy-n,n-diethylbenzamide, unii-hg05mo88tl, hg05mo88tl, acmc-20ak91, 2-hydroxy-n,n-diethylbenzamide | |
CCN(CC)C(=O)C1=CC=CC=C1O | |
193.246 | |
193.25 | |
N,N-Diethylsalicylamide |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 242-955-8
RTECSNumber : VN7320000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only