Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N'-Diethyloxamide, 98%

$173.28 - $173.28

Chemical Identifiers

Chemical Name or Material N,N'-Diethyloxamide
CAS 615-84-9
Molecular Formula C6H12N2O2
Formula Weight 144.17
MDL Number MFCD00043650
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2416722

  SDS
alfa aesar™
B2416722
100g Each for $173.28
Description & Specifications

Specifications

Formula Weight 144.17
Chemical Name or Material N,N'-Diethyloxamide
Melting Point 178° to 182°C
Assay 98%
Quantity 100g
Molecular Formula C6H12N2O2
Odor Odorless
MDL Number MFCD00043650
CAS 615-84-9