Learn More
N,N-Diethylnicotinamide, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC114470250
Specifications
N, N-Diethylnicotinamide | |
>112°C | |
Glass bottle | |
1.06 | |
Colorless to Yellow | |
97% | |
96.0 | |
96% min. (GC) | |
MFCD00006386 | |
15, 6627 | |
Solubility in water: miscible. Other solubilities: miscible with methanol,ethanol,acetone,chlorofo,rm,ether | |
CCN(CC)C(=O)C1=CN=CC=C1 | |
178.23 | |
178.23 | |
97% |
1.0600g/mL | |
Authentic | |
1.5240 to 1.526 | |
296°C to 300°C | |
25g | |
59-26-7 | |
100.0 | |
C10H14N2O | |
22, II, 34 | |
anacardone, anacordone, carbamidal, coramine, corvin, corvotone, n,n-diethylnicotinamide, nicamide, nicethamide, nikethamide | |
NCYVXEGFNDZQCU-UHFFFAOYSA-N | |
N,N-diethylpyridine-3-carboxamide | |
5497 | |
Liquid |
Chemical Identifiers
59-26-7 | |
178.23 | |
NCYVXEGFNDZQCU-UHFFFAOYSA-N | |
5497 | |
CCN(CC)C(=O)C1=CN=CC=C1 |
C10H14N2O | |
MFCD00006386 | |
anacardone, anacordone, carbamidal, coramine, corvin, corvotone, n,n-diethylnicotinamide, nicamide, nicethamide, nikethamide | |
N,N-diethylpyridine-3-carboxamide |
Safety and Handling
GHS H Statement:
Toxic in contact with skin.
Toxic if swallowed.
Toxic if inhaled.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 200-418-5
RTECSNumber : QS4375000
Recommended Storage : Product may darken on storage