Learn More
N,N-Diethylhydroxylamine, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 114421000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N, N-Diethylhydroxylamine | |
3710-84-7 | |
100.0 | |
0.8670g/mL | |
45°C | |
96% min. (GC) | |
C4H11NO | |
(C2H5)2NOH | |
100 g | |
06,187 | |
diethylhydroxylamine, ethanamine, n-ethyl-n-hydroxy, n-hydroxydiethylamine, n,n-diethylhydroxyamine, hydroxylamine, n,n-diethyl, unii-314i05edvh, ccris 964, hydroxydiethylamine, diethyl hydroxylamine, diethyl hydroxykamine | |
FVCOIAYSJZGECG-UHFFFAOYSA-N | |
N,N-diethylhydroxylamine | |
19463 | |
97% |
97% | |
96.0 | |
-26°C to 25°C | |
125.0°C to 130.0°C | |
Authentic | |
Glass bottle | |
1.4190 to 1.4220 | |
MFCD00002126 | |
04, 536 | |
0.867 | |
Solubility in water: soluble. | |
CCN(CC)O | |
89.14 | |
89.14 | |
Liquid |
Chemical Identifiers
3710-84-7 | |
89.14 | |
FVCOIAYSJZGECG-UHFFFAOYSA-N | |
19463 | |
CCN(CC)O |
C4H11NO | |
MFCD00002126 | |
diethylhydroxylamine, ethanamine, n-ethyl-n-hydroxy, n-hydroxydiethylamine, n,n-diethylhydroxyamine, hydroxylamine, n,n-diethyl, unii-314i05edvh, ccris 964, hydroxydiethylamine, diethyl hydroxylamine, diethyl hydroxykamine | |
N,N-diethylhydroxylamine |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fu
GHS Signal Word: Warning
EINECSNumber : 223-055-4
RTECSNumber : NC3500000
TSCA : TSCA
RUO – Research Use Only