Please login to your online account to display your discounted pricing

N,N'-Diethylethylenediamine, 95%, Acros Organics

$56.49 - $56.49

Chemical Identifiers

Chemical Name or Material N, N'-Diethylethylenediamine
Assay Percent Range 95%
Molecular Formula C6H16N2
Formula Weight 116.21
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC155520050

 
thermo fisher scientific
AC155520050
Glass bottle 5g Each for $56.49

AC155520250

 
thermo fisher scientific
AC155520250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 116.21
Quantity 5g
Chemical Name or Material N, N'-Diethylethylenediamine
CAS 111-74-0
Molecular Formula C6H16N2
Packaging Glass bottle
Assay Percent Range 95%