missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N,-Diethylchloroacetamide, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 351850050

 View more versions of this product

Catalog No. AC351850050


 

Specifications Description & Specifications

Specifications

≥97.5% (GC)
149.62
Clear colorless to light yellow
100.0
Glass bottle
C6H12ClNO
2315-36-8
MFCD00000928
Liquid or Solid
5g
1.0890g/mL
1.4680 to 1.4720 (20°C, 589nm)
N, N, -Diethylchloroacetamide
98%
97.5
Authentic

Chemical Identifiers

2315-36-8
100.0
C6H12ClNO
MFCD00000928
≥97.5% (GC)
97.5
98%
149.62
N, N, -Diethylchloroacetamide

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning