missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Diethylbenzamide, 99%

$92.74 - $282.73

Chemical Identifiers

CAS 1696-17-9
Molecular Formula C11H15NO
Formula Weight 177.25
MDL Number MFCD00026726
Beilstein 1909505
Chemical Name or Material N,N-Diethylbenzamide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0842714 SDS Alfa Aesar™
L0842714
25g Each for $92.74
AAL0842722 SDS Alfa Aesar™
L0842722
100g Each for $282.73
Specifications Description & Specifications

Chemical Identifiers

1696-17-9
177.25
1909505
C11H15NO
MFCD00026726
N,N-Diethylbenzamide

Specifications

1696-17-9
146° to 150°C (15mmHg)
>100°C (212°F)
177.25
1909505
99%
28° to 32°C
25g
C11H15NO
MFCD00026726
N,N-Diethylbenzamide

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : CV4202000

TSCA : TSCA