missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Diethyl-p-phenylenediamine sulfate, 97%

$99.14 - $357.11

Chemical Identifiers

CAS 6283-63-2
Molecular Formula C10H16N2·H2SO4
Formula Weight 262.33
MDL Number MFCD00012993
Beilstein 4219768
Synonym 4-Amino-N,N-diethylaniline sulfate
Chemical Name or Material N,N-Diethyl-p-phenylenediamine sulfate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA178320B SDS Alfa Aesar™
A178320B
1000g Each for $357.11
AAA1783230 SDS Alfa Aesar™
A1783230
250g Each for $99.14
AAA1783218 SDS Alfa Aesar™
A1783218
50g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Applications
N,N-Diethyl-p-phenylenediamine sulfate is used for the spectrophotometric determination of drugs such as salbutamol sulphate, ritodrine hydrochloride and isoxsuprine hydrochloride containing phenolic group. It is also used in the analysis of drugs like dapsone hydrochloride, sulfamethoxazole and sulfadiazine containing aromatic amino group. Further, it is used as a color developer and film developer in photography.

Solubility
Soluble in water and alcohol.

Notes
Light sensitive and air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Description & Specifications

Chemical Identifiers

6283-63-2
262.33
4219768
N,N-Diethyl-p-phenylenediamine sulfate
C10H16N2·H2SO4
MFCD00012993
4-Amino-N,N-diethylaniline sulfate

Specifications

6283-63-2
1000g
262.33
4219768
N,N-Diethyl-p-phenylenediamine sulfate
183° to 188°C
C10H16N2·H2SO4
MFCD00012993
4-Amino-N,N-diethylaniline sulfate
97%

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : SS9625000