missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N-Diethyl-p-phenylenediamine, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC407395000

 View more versions of this product

Catalog No. AC407395000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

N, N-Diethyl-p-phenylenediamine
139°C
Glass bottle
0.988
Undesignated
500g
97.5
97.5% min. (GC)
(CH3CH2)2NC6H4NH2
4-amino-n,n-diethylaniline, 4-diethylamino aniline, diethylaminoaniline, n,n-diethyl-1,4-benzenediamine, n,n-diethyl-1,4-phenylenediamine, n,n-diethyl-p-phenylenediamine, n1,n1-diethylbenzene-1,4-diamine, p-amino-n,n-diethylaniline, p-aminodiethylaniline, p-diethylamino aniline
QNGVNLMMEQUVQK-UHFFFAOYSA-N
4-N,4-N-diethylbenzene-1,4-diamine
7120
Liquid or Low Melting Solid
0.9880g/mL
Authentic
1.5690 to 1.5730 (20°C, 589nm)
268.0°C
23.0°C
93-05-0
100.0
C10H16N2
MFCD00007861
Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
CCN(CC)C1=CC=C(C=C1)N
164.25
164.25
98%

Chemical Identifiers

93-05-0
164.25
QNGVNLMMEQUVQK-UHFFFAOYSA-N
7120
CCN(CC)C1=CC=C(C=C1)N
C10H16N2
MFCD00007861
4-amino-n,n-diethylaniline, 4-diethylamino aniline, diethylaminoaniline, n,n-diethyl-1,4-benzenediamine, n,n-diethyl-1,4-phenylenediamine, n,n-diethyl-p-phenylenediamine, n1,n1-diethylbenzene-1,4-diamine, p-amino-n,n-diethylaniline, p-aminodiethylaniline, p-diethylamino aniline
4-N,4-N-diethylbenzene-1,4-diamine
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
Wear eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 202-214-1

SDS Documents