missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N,N-Diethyl-p-phenylenediamine, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 407390050
Specifications
N, N-Diethyl-p-phenylenediamine | |
139°C | |
Glass bottle | |
0.988 | |
Undesignated | |
5g | |
97.5 | |
97.5% min. (GC) | |
(CH3CH2)2NC6H4NH2 | |
4-amino-n,n-diethylaniline, 4-diethylamino aniline, diethylaminoaniline, n,n-diethyl-1,4-benzenediamine, n,n-diethyl-1,4-phenylenediamine, n,n-diethyl-p-phenylenediamine, n1,n1-diethylbenzene-1,4-diamine, p-amino-n,n-diethylaniline, p-aminodiethylaniline, p-diethylamino aniline | |
QNGVNLMMEQUVQK-UHFFFAOYSA-N | |
4-N,4-N-diethylbenzene-1,4-diamine | |
7120 | |
Liquid or Low Melting Solid |
0.9880g/mL | |
Authentic | |
1.5690 to 1.5730 (20°C, 589nm) | |
268.0°C | |
23.0°C | |
93-05-0 | |
100.0 | |
C10H16N2 | |
MFCD00007861 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether | |
CCN(CC)C1=CC=C(C=C1)N | |
164.25 | |
164.25 | |
98% |
Chemical Identifiers
93-05-0 | |
164.25 | |
QNGVNLMMEQUVQK-UHFFFAOYSA-N | |
7120 | |
CCN(CC)C1=CC=C(C=C1)N |
C10H16N2 | |
MFCD00007861 | |
4-amino-n,n-diethylaniline, 4-diethylamino aniline, diethylaminoaniline, n,n-diethyl-1,4-benzenediamine, n,n-diethyl-1,4-phenylenediamine, n,n-diethyl-p-phenylenediamine, n1,n1-diethylbenzene-1,4-diamine, p-amino-n,n-diethylaniline, p-aminodiethylaniline, p-diethylamino aniline | |
4-N,4-N-diethylbenzene-1,4-diamine |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement
Wear eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 202-214-1