Please login to your online account to display your discounted pricing

N,N-Diethyl-p-phenylenediamine, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  407390250

 View more versions of this product

Catalog No. AC407390250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Color Clear yellow-brown to dark brown
Formula Weight 164.25
Physical Form Liquid or Low Melting Solid
Quantity 25g
Density 0.988g/mL
CAS Max % 100.0
Synonym p-Aminodiethylaniline
Chemical Name or Material N, N-Diethyl-p-phenylenediamine
Packaging Glass bottle
CAS 93-05-0
Assay Percent Range 98%
Molecular Formula C10H16N2
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Causes severe skin burns and eye damage.
CAS Min % 97.5
MDL Number MFCD00007861
Refractive Index 1.5690 to 1.5730 (20°C, 589nm)
Infrared Spectrum Authentic
Linear Formula (CH3CH2)2NC6H4NH2

Specifications

Chemical Name or Material N, N-Diethyl-p-phenylenediamine
CAS 93-05-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H16N2
Linear Formula (CH3CH2)2NC6H4NH2
Formula Weight 164.25
MDL Number MFCD00007861
Synonym p-Aminodiethylaniline
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Causes severe skin burns and eye damage.

SDS & Safety Documents