Please login to your online account to display your discounted pricing

N,N-Diethyl-N'-methylethylenediamine, 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC314800250

 View more versions of this product

Catalog No. AC314800250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 130.24
Physical Form Liquid
Quantity 25g
Color Clear colorless
Density 0.8000g/mL
CAS Min % 96.0
CAS Max % 100.0
Molecular Formula C7H18N2
Chemical Name or Material N, N-Diethyl-N'-methylethylenediamine
Assay Percent Range 97%
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Name Note 97%
Refractive Index 1.4286 to 1.4306 (20°C, 589nm)
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
MDL Number MFCD00009053
Linear Formula (C2H5)2NCH2CH2NHCH3
CAS 104-79-0
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material N, N-Diethyl-N'-methylethylenediamine
Name Note 97%
CAS 104-79-0
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H18N2
Linear Formula (C2H5)2NCH2CH2NHCH3
Formula Weight 130.24
MDL Number MFCD00009053
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

SDS & Safety Documents