missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N-Diethyl-N'-methylethylenediamine, 97%, ACROS Organics™

Chemical Identifiers

CAS 104-79-0
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C7H18N2
Linear Formula (C2H5)2NCH2CH2NHCH3
Formula Weight 130.24
MDL Number MFCD00009053
Chemical Name or Material N, N-Diethyl-N'-methylethylenediamine, 97%
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC314800250 SDS Thermo Fisher Scientific
AC314800250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

104-79-0
100.0
C7H18N2
130.24
N, N-Diethyl-N'-methylethylenediamine, 97%
96.0
97%
(C2H5)2NCH2CH2NHCH3
MFCD00009053
≥96.0%

Specifications

Clear colorless
104-79-0
100.0
Authentic
1.4286 to 1.4306 (20°C, 589nm)
97%
(C2H5)2NCH2CH2NHCH3
MFCD00009053
≥96.0%
25g
96.0
0.8000g/mL
Glass bottle
Liquid
C7H18N2
130.24
N, N-Diethyl-N'-methylethylenediamine, 97%

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger