missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals N,N-Diethyl-m-toluamide, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 114575000

 View more versions of this product

Catalog No. AC114575000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N, N-Diethyl-m-toluamide
134-62-3
100.0
Undesignated
155°C
Authentic
CH3C6H4CON(C2H5)2
09, II, 325
15, 2859
500g
Solubility in water: negligible. Other solubilities: soluble in ethanol,chloroform,diethyl ether,,benzene
MMOXZBCLCQITDF-UHFFFAOYSA-N
N,N-diethyl-3-methylbenzamide
4284
191.27
98%
0.3% max.
98%
97.5
0.9955g/mL
111°C (1.0mmHg)
97.5% min. (GC)
C12H17NO
MFCD00009046
Glass bottle
1.5200 to 1.524
deet, n,n-diethyl-m-toluamide, diethyltoluamide, dieltamid, metadelphene, flypel, delphene, deta, detamide, autan
0.9955
CCN(CC)C(=O)C1=CC(=CC=C1)C
191.27
CHEBI:7071
Liquid
13.3 mPa.s (30°C)

Chemical Identifiers

134-62-3
191.27
MMOXZBCLCQITDF-UHFFFAOYSA-N
4284
N,N-diethyl-3-methylbenzamide
C12H17NO
MFCD00009046
deet, n,n-diethyl-m-toluamide, diethyltoluamide, dieltamid, metadelphene, flypel, delphene, deta, detamide, autan
CHEBI:7071
CCN(CC)C(=O)C1=CC(=CC=C1)C
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.

GHS P Statement:
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 205-149-7

RTECSNumber : XS3675000

TSCA : TSCA

SDS Documents

Documents

Product Certifications
Promotions

Promotions

RUO – Research Use Only