missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Diethyl-4-methylbenzenesulfonamide, 97%

Manufacturer:  Alfa Aesar™ H5505306

 View more versions of this product

Catalog No. AAH5505306



Specifications

Specifications

N,N-Diethyl-4-methylbenzenesulfonamide
60°C
649-15-0
MFCD00059369
AOJBACHWNDMRQP-UHFFFAOYSA-N
N,N-diethyl-4-methylbenzenesulfonamide
69539
Crystalline
White
5g
C11H17NO2S
acmc-209nol
CCN(CC)S(=O)(=O)C1=CC=C(C=C1)C
227.322
227.32
97%

Chemical Identifiers

649-15-0
227.322
AOJBACHWNDMRQP-UHFFFAOYSA-N
69539
CCN(CC)S(=O)(=O)C1=CC=C(C=C1)C
C11H17NO2S
MFCD00059369
acmc-209nol
N,N-diethyl-4-methylbenzenesulfonamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

RTECSNumber : XT5619800

TSCA : No

Recommended Storage : Ambient temperatures

Documents