missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Diethyl-2-methoxybenzamide, 97%

$85.14 - $269.86

Chemical Identifiers

CAS 51674-10-3
Molecular Formula C12H17NO2
Molecular Weight (g/mol) 207.273
MDL Number MFCD01010706
InChI Key ZFIIUAIWXACNKN-UHFFFAOYSA-N
Synonym n,n-diethyl-2-methoxy-benzamide, 2-methoxy-n,n-diethylbenzamide, 2-methoxy-n,n-diethyl-benzamide, benzamide,n,n-diethyl-2-methoxy, n,n-diethyl 2-methoxyphenyl carboxamide
PubChem CID 346975
IUPAC Name N,N-diethyl-2-methoxybenzamide
SMILES CCN(CC)C(=O)C1=CC=CC=C1OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH59793MD View Documents Alfa Aesar™
H59793MD
250mg Each for $85.14
 
AAH5979303 View Documents Alfa Aesar™
H5979303
1g Each for $269.86
 
Specifications

Chemical Identifiers

51674-10-3
207.273
ZFIIUAIWXACNKN-UHFFFAOYSA-N
346975
CCN(CC)C(=O)C1=CC=CC=C1OC
C12H17NO2
MFCD01010706
n,n-diethyl-2-methoxy-benzamide, 2-methoxy-n,n-diethylbenzamide, 2-methoxy-n,n-diethyl-benzamide, benzamide,n,n-diethyl-2-methoxy, n,n-diethyl 2-methoxyphenyl carboxamide
N,N-diethyl-2-methoxybenzamide

Specifications

250mg
C12H17NO2
n,n-diethyl-2-methoxy-benzamide, 2-methoxy-n,n-diethylbenzamide, 2-methoxy-n,n-diethyl-benzamide, benzamide,n,n-diethyl-2-methoxy, n,n-diethyl 2-methoxyphenyl carboxamide
CCN(CC)C(=O)C1=CC=CC=C1OC
207.273
207.27
N,N-Diethyl-2-methoxybenzamide
51674-10-3
MFCD01010706
ZFIIUAIWXACNKN-UHFFFAOYSA-N
N,N-diethyl-2-methoxybenzamide
346975
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning