missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Diethyl-2-fluoro-4-nitroaniline, 97%

$35.93 - $422.54

Chemical Identifiers

CAS 2689-37-4
Molecular Formula C10H13FN2O2
Molecular Weight (g/mol) 212.224
InChI Key FPVHBAPCTSCZKR-UHFFFAOYSA-N
Synonym Diethyl-(2-fluoro-4-nitrophenyl)amine
PubChem CID 67995604
IUPAC Name N,N-diethyl-2-fluoro-4-nitroaniline
SMILES CCN(CC)C1=C(C=C(C=C1)[N+](=O)[O-])F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63989MD View Documents Alfa Aesar™
H63989MD
250mg Each for $35.93
 
AAH6398903 View Documents Alfa Aesar™
H6398903
1g Each for $106.26
 
AAH6398906 View Documents Alfa Aesar™
H6398906
5g Each for $422.54
 
Specifications

Chemical Identifiers

2689-37-4
212.224
Diethyl-(2-fluoro-4-nitrophenyl)amine
N,N-diethyl-2-fluoro-4-nitroaniline
C10H13FN2O2
FPVHBAPCTSCZKR-UHFFFAOYSA-N
67995604
CCN(CC)C1=C(C=C(C=C1)[N+](=O)[O-])F

Specifications

250mg
C10H13FN2O2
2123723
FPVHBAPCTSCZKR-UHFFFAOYSA-N
N,N-diethyl-2-fluoro-4-nitroaniline
67995604
Solid
N,N-Diethyl-2-fluoro-4-nitroaniline
2689-37-4
UN2811
Diethyl-(2-fluoro-4-nitrophenyl)amine
CCN(CC)C1=C(C=C(C=C1)[N+](=O)[O-])F
212.224
212.22
97%
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.