missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Diethyl(2-chloro-1,1,2-trifluoroethyl)amine, 90%

$59.51 - $564.42

Chemical Identifiers

CAS 357-83-5
Molecular Formula C6H11ClF3N
Molecular Weight (g/mol) 189.606
MDL Number MFCD00054672
InChI Key BDZHKUAKSMWSAJ-UHFFFAOYSA-N
Synonym 2-chloro-1,1,2-trifluoroethyl diethylamine, n,n-diethyl 2-chloro-1,1,2-trifluoroethyl amine, n-2-chloro-1,1,2-trifluoroethyl diethylamine, yarovenko's reagent, acmc-20appo, 2-chloro-1,1,2-trifluorotriethylamine, 1,1,2-trifluoro-2-chloroethyldiethylamine, 2-chloro-1,1,2-trifluoroethyldiethylamine, ethanamine, 2-chloro-n,n-diethyltrifluoro, 1-diethylamino-1,1,2trifluoro-2-chloroethane
PubChem CID 136184
IUPAC Name 2-chloro-N,N-diethyl-1,1,2-trifluoroethanamine
SMILES CCN(CC)C(C(F)Cl)(F)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1733006 View Documents Alfa Aesar™
L1733006
5g Each for $59.51
 
AAL1733014 View Documents Alfa Aesar™
L1733014
25g Each for $195.85
 
AAL1733022 View Documents Alfa Aesar™
L1733022
100g Each for $564.42
 
Specifications

Chemical Identifiers

357-83-5
189.606
BDZHKUAKSMWSAJ-UHFFFAOYSA-N
136184
CCN(CC)C(C(F)Cl)(F)F
C6H11ClF3N
MFCD00054672
2-chloro-1,1,2-trifluoroethyl diethylamine, n,n-diethyl 2-chloro-1,1,2-trifluoroethyl amine, n-2-chloro-1,1,2-trifluoroethyl diethylamine, yarovenko's reagent, acmc-20appo, 2-chloro-1,1,2-trifluorotriethylamine, 1,1,2-trifluoro-2-chloroethyldiethylamine, 2-chloro-1,1,2-trifluoroethyldiethylamine, ethanamine, 2-chloro-n,n-diethyltrifluoro, 1-diethylamino-1,1,2trifluoro-2-chloroethane
2-chloro-N,N-diethyl-1,1,2-trifluoroethanamine

Specifications

38°C (10mmHg)
357-83-5
ClFCHCF2N(CH2CH3)2
UN3267
2-chloro-1,1,2-trifluoroethyl diethylamine, n,n-diethyl 2-chloro-1,1,2-trifluoroethyl amine, n-2-chloro-1,1,2-trifluoroethyl diethylamine, yarovenko's reagent, acmc-20appo, 2-chloro-1,1,2-trifluorotriethylamine, 1,1,2-trifluoro-2-chloroethyldiethylamine, 2-chloro-1,1,2-trifluoroethyldiethylamine, ethanamine, 2-chloro-n,n-diethyltrifluoro, 1-diethylamino-1,1,2trifluoro-2-chloroethane
CCN(CC)C(C(F)Cl)(F)F
189.606
189.61
N,N-Diethyl(2-chloro-1,1,2-trifluoroethyl)amine
5g
C6H11ClF3N
MFCD00054672
1098675
BDZHKUAKSMWSAJ-UHFFFAOYSA-N
2-chloro-N,N-diethyl-1,1,2-trifluoroethanamine
136184
90%
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.

Recommended Storage : 2° to 8°C