missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Diethyl-2,5-difluorobenzamide, 97%

Manufacturer:  Alfa Aesar™ H61556MD

 View more versions of this product

Catalog No. AAH61556MD



Specifications

Specifications

N,N-Diethyl-2,5-difluorobenzamide
131401-55-3
MFCD00134465
JEEJMEMWBHLCKZ-UHFFFAOYSA-N
N,N-diethyl-2,5-difluorobenzamide
5044848
97%
250mg
C11H13F2NO
2,5-difluoro-n,n-diethylbenzamide
CCN(CC)C(=O)C1=C(C=CC(=C1)F)F
213.228
213.23

Chemical Identifiers

131401-55-3
213.228
JEEJMEMWBHLCKZ-UHFFFAOYSA-N
5044848
CCN(CC)C(=O)C1=C(C=CC(=C1)F)F
C11H13F2NO
MFCD00134465
2,5-difluoro-n,n-diethylbenzamide
N,N-diethyl-2,5-difluorobenzamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents