missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Diethyl-2,3,3,3-tetrafluoropropionamide, 95%

$80.84 - $197.69

Chemical Identifiers

CAS 392-63-2
Molecular Formula C7H11F4NO
Molecular Weight (g/mol) 201.165
MDL Number MFCD00069160
InChI Key RQUXJBKONYSFAU-UHFFFAOYSA-N
Synonym acmc-20ak99
PubChem CID 2774042
IUPAC Name N,N-diethyl-2,3,3,3-tetrafluoropropanamide
SMILES CCN(CC)C(=O)C(C(F)(F)F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1673914 View Documents Alfa Aesar™
L1673914
25g Each for $80.84
 
AAL1673922 View Documents Alfa Aesar™
L1673922
100g Each for $197.69
 
Specifications

Chemical Identifiers

392-63-2
201.165
RQUXJBKONYSFAU-UHFFFAOYSA-N
2774042
CCN(CC)C(=O)C(C(F)(F)F)F
C7H11F4NO
MFCD00069160
acmc-20ak99
N,N-diethyl-2,3,3,3-tetrafluoropropanamide

Specifications

1.228
58°C (5mmHg)
392-63-2
CF3CHFCON(CH2CH3)2
1780406
RQUXJBKONYSFAU-UHFFFAOYSA-N
N,N-diethyl-2,3,3,3-tetrafluoropropanamide
2774042
95%
1.392
25g
C7H11F4NO
MFCD00069160
acmc-20ak99
CCN(CC)C(=O)C(C(F)(F)F)F
201.165
201.16
N,N-Diethyl-2,3,3,3-tetrafluoropropionamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

TSCA : No

Recommended Storage : Ambient temperatures