missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Diethyl-2,3,3,3-tetrafluoropropionamide, 95%

Manufacturer:  Alfa Aesar™ L1673922

 View more versions of this product

Catalog No. AAL1673922



Specifications

Specifications

N,N-Diethyl-2,3,3,3-tetrafluoropropionamide
100g
C7H11F4NO
MFCD00069160
acmc-20ak99
CCN(CC)C(=O)C(C(F)(F)F)F
201.165
201.16
58°C (5mmHg)
392-63-2
CF3CHFCON(CH2CH3)2
1780406
RQUXJBKONYSFAU-UHFFFAOYSA-N
N,N-diethyl-2,3,3,3-tetrafluoropropanamide
2774042
95%

Chemical Identifiers

392-63-2
201.165
RQUXJBKONYSFAU-UHFFFAOYSA-N
2774042
CCN(CC)C(=O)C(C(F)(F)F)F
C7H11F4NO
MFCD00069160
acmc-20ak99
N,N-diethyl-2,3,3,3-tetrafluoropropanamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

Documents