Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Diethyl-2,2,2-trifluoroacetamide, 97%

Chemical Identifiers

Chemical Name or Material N,N-Diethyl-2,2,2-trifluoroacetamide
CAS 360-92-9
Molecular Formula C6H10F3NO
Formula Weight 169.15
MDL Number MFCD00192034
UN Number UN1993
Synonym Trifluoroacetic acid diethylamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2582906

  SDS
alfa aesar™
H2582906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2582903

  SDS
alfa aesar™
H2582903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 169.15
Synonym Trifluoroacetic acid diethylamide
MDL Number MFCD00192034
Molecular Formula C6H10F3NO
Health Hazard 2 GHS H Statement
H226-H302-H312-H332-H315-H319-H335
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Boiling Point 157° to 160°C
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Health Hazard 1 Warning
Flash Point 43°C (110°F)
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
UN Number UN1993
CAS 360-92-9
Chemical Name or Material N,N-Diethyl-2,2,2-trifluoroacetamide
Assay 97%
Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H302-H312-H332-H315-H319-H335
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.