missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Diethyl-1,3-propanediamine, 99%

Manufacturer:  Alfa Aesar™ L13888AE

 View more versions of this product

Catalog No. AAL13888AE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Applications
3-Diethylaminopropylamine is used as a solvent, extraction agents and organic synthesis intermediates, dyes, pigments, surface active agent.

Solubility
Fully miscible in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
Specifications

Specifications

N,N-Diethyl-1,3-propanediamine
168°C to 171°C
100mL
C7H18N2
MFCD00008218
741879
QOHMWDJIBGVPIF-UHFFFAOYSA-N
N',N'-diethylpropane-1,3-diamine
61011
99%
51°C (124°F)
-60°C
104-78-9
(CH3CH2)2N(CH2)3NH2
UN2684
3-diethylaminopropylamine, 3-diethylamino propylamine, n,n-diethyl-1,3-propanediamine, 3-aminopropyl diethylamine, n1,n1-diethylpropane-1,3-diamine, 1,3-propanediamine, n,n-diethyl, diethylaminotrimethylenamine, n,n-diethyl-1,3-diaminopropane, 1-diethylamino propylamine-3, n-3-diethylaminopropyl amine
CCN(CC)CCCN
130.235
130.23

Chemical Identifiers

104-78-9
130.235
QOHMWDJIBGVPIF-UHFFFAOYSA-N
61011
CCN(CC)CCCN
C7H18N2
MFCD00008218
3-diethylaminopropylamine, 3-diethylamino propylamine, n,n-diethyl-1,3-propanediamine, 3-aminopropyl diethylamine, n1,n1-diethylpropane-1,3-diamine, 1,3-propanediamine, n,n-diethyl, diethylaminotrimethylenamine, n,n-diethyl-1,3-diaminopropane, 1-diethylamino propylamine-3, n-3-diethylaminopropyl amine
N',N'-diethylpropane-1,3-diamine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H226-H302-H312-H317
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: 3-DIETHYAMINO-PROPYLAMINE

RTECSNumber : TX7350000

TSCA : TSCA

Documents