Please login to your online account to display your discounted pricing

N,N-Diethyl-1,1,1-trimethylsilylamine, 98+%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  155690250

 View more versions of this product

Catalog No. AC155690250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥98.0%
Formula Weight 145.32
Quantity 25g
Color Clear colorless to yellow
EINECS Number 213-637-6
TSCA TSCA
Density 0.76g/mL
Fieser 03,317; 04,544; 06,634
CAS Min % 98.0
CAS Max % 100.0
Refractive Index 1.4103 to 1.4123 (20°C, 589nm)
Synonym TMSDEA
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Highly flammable liquid and vapor.
Packaging Glass bottle
Chemical Name or Material N,N-Diethyl-1,1,1-trimethylsilylamine
Name Note 98+%
Assay Percent Range 98+%
Molecular Formula C7H19NSi
Linear Formula (C2H5)2NSi(CH3)3
MDL Number MFCD00009040
CAS 996-50-9
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material N,N-Diethyl-1,1,1-trimethylsilylamine
Name Note 98+%
CAS 996-50-9
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C7H19NSi
Linear Formula (C2H5)2NSi(CH3)3
Formula Weight 145.32
MDL Number MFCD00009040
Synonym TMSDEA
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Highly flammable liquid and vapor.

SDS & Safety Documents