Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N'-Dicyclohexylthiourea, 98+%

$45.81 - $121.74

Chemical Identifiers

Chemical Name or Material N,N'-Dicyclohexylthiourea
CAS 1212-29-9
Molecular Formula C13H24N2S
Formula Weight 240.41
MDL Number MFCD00021316
Beilstein 517587
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0778514

  SDS
alfa aesar™
L0778514
25g Each for $45.81

AAL0778522

  SDS
alfa aesar™
L0778522
100g Each for $121.74
Description & Specifications

Specifications

Formula Weight 240.41
Chemical Name or Material N,N'-Dicyclohexylthiourea
TSCA TSCA
Melting Point 180° to 182°C
Assay ≥98%
Quantity 25g
Molecular Formula C13H24N2S
Odor Odorless
MDL Number MFCD00021316
CAS 1212-29-9
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P280f
Wear protective clothing.
RTECS Number YS9300000
Beilstein 517587
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Safety and Handling

Warning

GHS P Statement
P280f
Wear protective clothing.

GHS H Statement
H302
Harmful if swallowed.