Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N'-Dicarbamoylhydrazine, 98%

$30.45 - $461.07

Chemical Identifiers

Chemical Name or Material N,N'-Dicarbamoylhydrazine
CAS 110-21-4
Molecular Formula H2NCONHNHCONH2
Formula Weight 118.1
MDL Number MFCD00025398
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1826009

  SDS
alfa aesar™
A1826009
10g Each for $30.45

AAA1826030

  SDS
alfa aesar™
A1826030
250g Each for $461.07

AAA1826018

  SDS
alfa aesar™
A1826018
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material N,N'-Dicarbamoylhydrazine
CAS 110-21-4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Melting Point 239° to 241°C (dec.)
Assay 98%
Quantity 10g
Molecular Formula H2NCONHNHCONH2
Formula Weight 118.1
MDL Number MFCD00025398
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.