Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Dibutylbenzenesulfonamide, 97%

$35.55 - $106.36

Chemical Identifiers

Chemical Name or Material N,N-Dibutylbenzenesulfonamide
CAS 5339-59-3
Molecular Formula C14H23NO2S
Formula Weight 269.41
MDL Number MFCD00151808
Beilstein 2852807
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1339403

  SDS
alfa aesar™
L1339403
1g Each for $35.55

AAL1339406

  SDS
alfa aesar™
L1339406
5g Each for $106.36
Description & Specifications

Specifications

Formula Weight 269.41
Chemical Name or Material N,N-Dibutylbenzenesulfonamide
CAS 5339-59-3
Assay 97%
Health Hazard 1 Warning
MDL Number MFCD00151808
Beilstein 2852807
Boiling Point 211° to 212°C (17mmHg)
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Molecular Formula C14H23NO2S
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.