missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Dibutylbenzenesulfonamide, 97%

Manufacturer:  Alfa Aesar™ L1339403

 View more versions of this product

Catalog No. AAL1339403



Specifications

Specifications

N,N-Dibutylbenzenesulfonamide
1g
C14H23NO2S
2852807
YKXINVWHNNYQMA-UHFFFAOYSA-N
N,N-dibutylbenzenesulfonamide
220596
97%
211°C to 212°C (17mmHg)
5339-59-3
MFCD00151808
acmc-20ak9e, dibutyl phenylsulfonyl amine, n,n-dibutylbenzenesulphonamide, benzenesulfonamide, n,n-dibutyl
CCCCN(CCCC)S(=O)(=O)C1=CC=CC=C1
269.403
269.41

Chemical Identifiers

5339-59-3
269.403
YKXINVWHNNYQMA-UHFFFAOYSA-N
220596
CCCCN(CCCC)S(=O)(=O)C1=CC=CC=C1
C14H23NO2S
MFCD00151808
acmc-20ak9e, dibutyl phenylsulfonyl amine, n,n-dibutylbenzenesulphonamide, benzenesulfonamide, n,n-dibutyl
N,N-dibutylbenzenesulfonamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents