Learn More
N,N-Dibutylaniline, 97%, Thermo Scientific Chemicals
$44.45 - $44.45
Chemical Identifiers
CAS | 613-29-6 |
---|---|
Molecular Formula | C14H23N |
Molecular Weight (g/mol) | 205.35 |
MDL Number | MFCD00035799 |
InChI Key | FZPXKEPZZOEPGX-UHFFFAOYSA-N |
Synonym | benzenamine, n,n-dibutyl, n,n-di-n-butylaniline, dibutylaniline, aniline, n,n-dibutyl, dibutyl-phenyl-amine, dibutylphenylamine, n,n-dibutyl aniline, n-phenyldibutylamine, n,n-dibutylbenzenamine, dibutylamine, n-phenyl |
PubChem CID | 61154 |
IUPAC Name | N,N-dibutylaniline |
SMILES | CCCCN(CCCC)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1339006
|
Thermo Scientific Chemicals
L1339006 |
5 g |
Each for $44.45
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
613-29-6 | |
205.35 | |
FZPXKEPZZOEPGX-UHFFFAOYSA-N | |
61154 | |
CCCCN(CCCC)C1=CC=CC=C1 |
C14H23N | |
MFCD00035799 | |
benzenamine, n,n-dibutyl, n,n-di-n-butylaniline, dibutylaniline, aniline, n,n-dibutyl, dibutyl-phenyl-amine, dibutylphenylamine, n,n-dibutyl aniline, n-phenyldibutylamine, n,n-dibutylbenzenamine, dibutylamine, n-phenyl | |
N,N-dibutylaniline |
Specifications
613-29-6 | |
269°C to 270°C | |
C14H23N | |
MFCD00035799 | |
UN2810 | |
benzenamine, n,n-dibutyl, n,n-di-n-butylaniline, dibutylaniline, aniline, n,n-dibutyl, dibutyl-phenyl-amine, dibutylphenylamine, n,n-dibutyl aniline, n-phenyldibutylamine, n,n-dibutylbenzenamine, dibutylamine, n-phenyl | |
CCCCN(CCCC)C1=CC=CC=C1 | |
205.35 | |
205.35 | |
N,N-Dibutylaniline |
0.906 | |
>110°C (230°F) | |
1.518 | |
5 g | |
2210422 | |
FZPXKEPZZOEPGX-UHFFFAOYSA-N | |
N,N-dibutylaniline | |
61154 | |
97% |
Safety and Handling
GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber : 210-335-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only