Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Dibutylaniline, 97%


Quantity 25g

Manufacturer: alfa aesar™  L1339014

 View more versions of this product

Catalog No. AAL1339014

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 205.35
Chemical Name or Material N,N-Dibutylaniline
Molecular Formula C14H23N
Assay 97%
Quantity 25g
Flash Point >110°C (230°F)
MDL Number MFCD00035799
CAS 613-29-6
UN Number UN2810
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Boiling Point 269° to 270°C
Beilstein 2210422
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

Specifications

Chemical Name or Material N,N-Dibutylaniline
CAS 613-29-6
Molecular Formula C14H23N
Formula Weight 205.35
MDL Number MFCD00035799
UN Number UN2810
Beilstein 2210422
Accessories

Danger

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.