Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N'-Dibenzylthiourea, 98%

$109.92 - $334.03

Chemical Identifiers

Chemical Name or Material N,N'-Dibenzylthiourea
CAS 1424-14-2
Molecular Formula C15H16N2S
Formula Weight 256.37
MDL Number MFCD00022013
Beilstein 989212
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0439014

  SDS
alfa aesar™
L0439014
25g Each for $109.92

AAL0439022

  SDS
alfa aesar™
L0439022
100g Each for $334.03
Description & Specifications

Specifications

Formula Weight 256.37
Chemical Name or Material N,N'-Dibenzylthiourea
MDL Number MFCD00022013
CAS 1424-14-2
Health Hazard 1 Warning
TSCA TSCA
Melting Point 146° to 149°C
Assay 98%
Quantity 25g
Molecular Formula C15H16N2S
Odor Odorless
Beilstein 989212
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P280f-P308+P313-P330-P501a
Wear protective clothing.
IF exposed or concerned: Get medical advice/attention.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P280f-P308+P313-P330-P501a
Wear protective clothing.
IF exposed or concerned: Get medical advice/attention.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.