missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N'-Dibenzyloxycarbonyl-L-cystine, 98%

Manufacturer:  Alfa Aesar™ H6391703

 View more versions of this product

Catalog No. AAH6391703



Specifications

Specifications

N,N'-Dibenzyloxycarbonyl-L-cystine
C22H24N2O8S2
2550640
PTRQEEVKHMDMCF-UHFFFAOYNA-N
(2R)-3-[[(2R)-2-carboxy-2-(phenylmethoxycarbonylamino)ethyl]disulfanyl]-2-(phenylmethoxycarbonylamino)propanoic acid
385972
Powder
White
1g
6968-11-2
MFCD00022030
z-cys-oh 2, cbz-cys-oh 2, di-z-l-cystine, n,n'-dicarbobenzoxy-l-cystine, n,n'-di-cbz-l-cystine, 2r-2-benzyloxy carbonyl amino-3-2r-2-benzyloxy carbonyl amino-2-carboxyethyl disulfanyl propanoic acid, n,n'-dibenzyloxycarbonyl-l-cystine, n,n'-bis benzyloxycarbonyl-l-cystine, ambotzzaa1190, pubchem19022
C1=CC=C(C=C1)COC(=O)NC(CSSCC(C(=O)O)NC(=O)OCC2=CC=CC=C2)C(=O)O
508.56
508.56
98%
119°C

Chemical Identifiers

6968-11-2
508.56
PTRQEEVKHMDMCF-UHFFFAOYNA-N
385972
C1=CC=C(C=C1)COC(=O)NC(CSSCC(C(=O)O)NC(=O)OCC2=CC=CC=C2)C(=O)O
C22H24N2O8S2
MFCD00022030
z-cys-oh 2, cbz-cys-oh 2, di-z-l-cystine, n,n'-dicarbobenzoxy-l-cystine, n,n'-di-cbz-l-cystine, 2r-2-benzyloxy carbonyl amino-3-2r-2-benzyloxy carbonyl amino-2-carboxyethyl disulfanyl propanoic acid, n,n'-dibenzyloxycarbonyl-l-cystine, n,n'-bis benzyloxycarbonyl-l-cystine, ambotzzaa1190, pubchem19022
(2R)-3-[[(2R)-2-carboxy-2-(phenylmethoxycarbonylamino)ethyl]disulfanyl]-2-(phenylmethoxycarbonylamino)propanoic acid
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

SDS Documents