Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N'-Dibenzylethylenediamine, 97%

$76.39 - $76.39

Chemical Identifiers

Chemical Name or Material N,N'-Dibenzylethylenediamine
CAS 140-28-3
Molecular Formula C16H20N2
Linear Formula C6H5CH2NHCH2CH2NHCH2C6H5
Formula Weight 240.35
MDL Number MFCD00004771
UN Number UN2735
Beilstein 786668
Merck Index 14,1064
Synonym Benzathine; 1,2-Bis(benzylamino)ethane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0354014

  SDS
alfa aesar™
L0354014
25g Each for $76.39

AAL0354022

  SDS
alfa aesar™
L0354022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 240.35
Chemical Name or Material N,N'-Dibenzylethylenediamine
MDL Number MFCD00004771
Health Hazard 2 GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
RTECS Number KV3325000
CAS 140-28-3
Synonym Benzathine; 1,2-Bis(benzylamino)ethane
Merck Index 14,1064
UN Number UN2735
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA TSCA
Health Hazard 1 Danger
Linear Formula C6H5CH2NHCH2CH2NHCH2C6H5
Assay 97%
Boiling Point 184° to 186°C (2mmHg)
Molecular Formula C16H20N2
Melting Point 23° to 25°C
Flash Point >110°C (230°F)
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 25g
Beilstein 786668

Applications
N,N′-Dibenzylethylenediamine is an intermediate of cephalosporin and penicillin.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.

Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.