Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Dibenzylaniline, 99%

$64.65 - $180.81

Chemical Identifiers

Chemical Name or Material N,N-Dibenzylaniline
CAS 91-73-6
Molecular Formula C20H19N
Formula Weight 273.37
MDL Number MFCD00022015
Synonym N-Phenyldibenzylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3209114

  SDS
alfa aesar™
H3209114
25g Each for $64.65

AAH3209122

  SDS
alfa aesar™
H3209122
100g Each for $180.81
Description & Specifications

Specifications

Formula Weight 273.37
Assay 99%
Quantity 25g
Molecular Formula C20H19N
Health Hazard 3 GHS P Statement
P280-P301+P310-P361-P312-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym N-Phenyldibenzylamine
TSCA TSCA
Health Hazard 2 GHS H Statement
H301-H311
Toxic if swallowed.
Toxic in contact with skin.
Melting Point 69° to 71°C
MDL Number MFCD00022015
Health Hazard 1 Danger
Boiling Point 180°C (1mmHg)
CAS 91-73-6
Chemical Name or Material N,N-Dibenzylaniline
Safety and Handling

Danger

GHS P Statement
P280-P301+P310-P361-P312-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311
Toxic if swallowed.
Toxic in contact with skin.