missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (+)-N,N'-Dibenzyl-L-tartaric diamide

$43.32 - $710.92

Chemical Identifiers

CAS 88393-56-0
Molecular Formula C18H20N2O4
Molecular Weight (g/mol) 328.368
MDL Number MFCD00064478
InChI Key BBYSAHVLSFBCMN-HZPDHXFCSA-N
Synonym 2r,3r-n1,n4-dibenzyl-2,3-dihydroxysuccinamide, 2r,3r-n,n'-dibenzyl-2,3-dihydroxysuccinamide, dibenzyl tartramide, n,n'-dibenzyl-l-tartramide, +-n,n-dibenzyl-l-tartaric diamide, +-n,n'-dibenzyl-l-tartaric diamide, 2r,3r-n,n'-dibenzyl-2,3-dihydroxybutanediamide, n,n inverted exclamation marka-dibenzyl-l-tartramide, +-n,n inverted exclamation marka-dibenzyl-l-tartaric diamide
PubChem CID 7271811
IUPAC Name (2R,3R)-N,N'-dibenzyl-2,3-dihydroxybutanediamide
SMILES C1=CC=C(C=C1)CNC(=O)C(C(C(=O)NCC2=CC=CC=C2)O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6573503 View Documents Alfa Aesar™
H6573503
1g Each for $43.32
 
AAH6573506 View Documents Alfa Aesar™
H6573506
5g Each for $172.72
 
AAH6573514 View Documents Alfa Aesar™
H6573514
25g Each for $710.92
 
Specifications

Chemical Identifiers

88393-56-0
328.368
BBYSAHVLSFBCMN-HZPDHXFCSA-N
7271811
C1=CC=C(C=C1)CNC(=O)C(C(C(=O)NCC2=CC=CC=C2)O)O
C18H20N2O4
MFCD00064478
2r,3r-n1,n4-dibenzyl-2,3-dihydroxysuccinamide, 2r,3r-n,n'-dibenzyl-2,3-dihydroxysuccinamide, dibenzyl tartramide, n,n'-dibenzyl-l-tartramide, +-n,n-dibenzyl-l-tartaric diamide, +-n,n'-dibenzyl-l-tartaric diamide, 2r,3r-n,n'-dibenzyl-2,3-dihydroxybutanediamide, n,n inverted exclamation marka-dibenzyl-l-tartramide, +-n,n inverted exclamation marka-dibenzyl-l-tartaric diamide
(2R,3R)-N,N'-dibenzyl-2,3-dihydroxybutanediamide

Specifications

White
1g
C18H20N2O4
4923000
BBYSAHVLSFBCMN-HZPDHXFCSA-N
(2R,3R)-N,N'-dibenzyl-2,3-dihydroxybutanediamide
7271811
Powder
198°C to 200°C
88393-56-0
MFCD00064478
2r,3r-n1,n4-dibenzyl-2,3-dihydroxysuccinamide, 2r,3r-n,n'-dibenzyl-2,3-dihydroxysuccinamide, dibenzyl tartramide, n,n'-dibenzyl-l-tartramide, +-n,n-dibenzyl-l-tartaric diamide, +-n,n'-dibenzyl-l-tartaric diamide, 2r,3r-n,n'-dibenzyl-2,3-dihydroxybutanediamide, n,n inverted exclamation marka-dibenzyl-l-tartramide, +-n,n inverted exclamation marka-dibenzyl-l-tartaric diamide
C1=CC=C(C=C1)CNC(=O)C(C(C(=O)NCC2=CC=CC=C2)O)O
328.368
328.37
(+)-N,N'-Dibenzyl-L-tartaric diamide