missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(+)-N,N'-Dibenzyl-L-tartaric diamide, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6573506
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
(+)-N,N'-Dibenzyl-L-tartaric diamide | |
198°C to 200°C | |
C18H20N2O4 | |
5 g | |
2r,3r-n1,n4-dibenzyl-2,3-dihydroxysuccinamide, 2r,3r-n,n'-dibenzyl-2,3-dihydroxysuccinamide, dibenzyl tartramide, n,n'-dibenzyl-l-tartramide, +-n,n-dibenzyl-l-tartaric diamide, +-n,n'-dibenzyl-l-tartaric diamide, 2r,3r-n,n'-dibenzyl-2,3-dihydroxybutanediamide, n,n inverted exclamation marka-dibenzyl-l-tartramide, +-n,n inverted exclamation marka-dibenzyl-l-tartaric diamide | |
O[C@H]([C@@H](O)C(=O)NCC1=CC=CC=C1)C(=O)NCC1=CC=CC=C1 | |
328.37 | |
328.37 |
88393-56-0 | |
White | |
MFCD00064478 | |
4923000 | |
BBYSAHVLSFBCMN-MNSHXRSINA-N | |
(2R,3R)-N,N'-dibenzyl-2,3-dihydroxybutanediamide | |
7271811 | |
Powder |
Chemical Identifiers
88393-56-0 | |
328.37 | |
BBYSAHVLSFBCMN-MNSHXRSINA-N | |
7271811 | |
O[C@H]([C@@H](O)C(=O)NCC1=CC=CC=C1)C(=O)NCC1=CC=CC=C1 |
C18H20N2O4 | |
MFCD00064478 | |
2r,3r-n1,n4-dibenzyl-2,3-dihydroxysuccinamide, 2r,3r-n,n'-dibenzyl-2,3-dihydroxysuccinamide, dibenzyl tartramide, n,n'-dibenzyl-l-tartramide, +-n,n-dibenzyl-l-tartaric diamide, +-n,n'-dibenzyl-l-tartaric diamide, 2r,3r-n,n'-dibenzyl-2,3-dihydroxybutanediamide, n,n inverted exclamation marka-dibenzyl-l-tartramide, +-n,n inverted exclamation marka-dibenzyl-l-tartaric diamide | |
(2R,3R)-N,N'-dibenzyl-2,3-dihydroxybutanediamide |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures