Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Dibenzyl-4-bromoaniline, 95%

$29.70 - $60.80

Chemical Identifiers

Chemical Name or Material N,N-Dibenzyl-4-bromoaniline
CAS 65145-14-4
Molecular Formula C20H18BrN
Formula Weight 352.28
MDL Number MFCD09037473
Synonym N-(4-Bromophenyl)dibenzylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2725103

  SDS
alfa aesar™
H2725103
1g Each for $60.80

AAH27251MD

  SDS
alfa aesar™
H27251MD
250mg Each for $29.70
Description & Specifications

Specifications

Formula Weight 352.28
Chemical Name or Material N,N-Dibenzyl-4-bromoaniline
Assay 95%
Quantity 1g
Molecular Formula C20H18BrN
MDL Number MFCD09037473
CAS 65145-14-4
Synonym N-(4-Bromophenyl)dibenzylamine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.