Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N'-Dibenzoylhydrazine, 98+%

$64.03 - $575.69

Chemical Identifiers

Chemical Name or Material N,N'-Dibenzoylhydrazine
CAS 787-84-8
Molecular Formula (C6H5)CONHNHCO(C6H5)
Formula Weight 240.26
MDL Number MFCD00003068
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2508106

 
alfa aesar™
B2508106
5g Each for $64.03

AAB2508114

 
alfa aesar™
B2508114
25g Each for $190.86

AAB2508122

 
alfa aesar™
B2508122
100g Each for $575.69
Description & Specifications

Specifications

Formula Weight 240.26
Chemical Name or Material N,N'-Dibenzoylhydrazine
TSCA TSCA
Melting Point 238° to 240°C
Assay ≥98%
Quantity 5g
Molecular Formula (C6H5)CONHNHCO(C6H5)
MDL Number MFCD00003068
CAS 787-84-8