missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N'-Diaminoguanidine monohydrochloride, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 162400050

 View more versions of this product

Catalog No. AC162400050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

N, N'-Diaminoguanidine monohydrochloride
Glass bottle
178°C to 182°C
98%
97.5
97.5% min. (Argentometry)
H2NNHC(=NH)NHNH2·HCl
03, 122
HAZRIBSLCUYMQP-UHFFFAOYSA-N
1,2-diaminoguanidine;hydrochloride
9566041
Granular Powder
Authentic
White to Beige
5g
36062-19-8
100.0
CH7N5·HCl
MFCD00012948
1,2-diaminoguanidine hydrochloride
C(=NN)(N)NN.Cl
125.56
125.56
98%

Chemical Identifiers

36062-19-8
125.56
HAZRIBSLCUYMQP-UHFFFAOYSA-N
9566041
C(=NN)(N)NN.Cl
CH7N5·HCl
MFCD00012948
1,2-diaminoguanidine hydrochloride
1,2-diaminoguanidine;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 252-854-0

SDS Documents