missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N'-Diallylthiourea, 96%

$78.99 - $302.18

Chemical Identifiers

CAS 6601-20-3
Molecular Formula C7H12N2S
Formula Weight 156.25
MDL Number MFCD00014945
Beilstein 1754800
Chemical Name or Material N,N'-Diallylthiourea
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0864806 SDS Alfa Aesar™
L0864806
5g Each for $78.99
AAL0864814 SDS Alfa Aesar™
L0864814
25g Each for $302.18
Specifications Description & Specifications

Chemical Identifiers

6601-20-3
156.25
1754800
C7H12N2S
MFCD00014945
N,N'-Diallylthiourea

Specifications

6601-20-3
5g
Odorless
MFCD00014945
N,N'-Diallylthiourea
42° to 45°C
C7H12N2S
156.25
1754800
96%

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P280f
Wear protective clothing.

Warning

TSCA : TSCA