Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N'-Diallyl-L-tartardiamide, Electrophoresis Reagent, 99%

Chemical Identifiers

Chemical Name or Material N,N'-Diallyl-L-tartardiamide
CAS 58477-85-3
Name Note Electrophoresis Reagent
Formula Weight 228.25
MDL Number MFCD00008640
Beilstein 1712934
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ6658718

 
alfa aesar™
J6658718
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ6658730

 
alfa aesar™
J6658730
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 184° to 186°C
Formula Weight 228.25
Chemical Name or Material N,N'-Diallyl-L-tartardiamide
MDL Number MFCD00008640
Physical Form Flakes
Assay 99%
TSCA TSCA
Color White to off-white
Name Note Electrophoresis Reagent
Beilstein 1712934
Quantity 50g
CAS 58477-85-3