missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N'-Diallyl-L-tartardiamide, 99%, ACROS Organics™

Chemical Identifiers

CAS 58477-85-3
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C10H16N2O4
Linear Formula [-CH(OH)CONHCH2CH=CH2]2
Formula Weight 228.25
MDL Number MFCD00008640
Synonym DATD
Chemical Name or Material N, N'-Diallyl-L-tartardiamide, 99%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC406330250 SDS Thermo Fisher Scientific
406330250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

58477-85-3
100.0
C10H16N2O4
228.25
DATD
≥98.5%
98.5
99%
[-CH(OH)CONHCH2CH=CH2]2
MFCD00008640
N, N'-Diallyl-L-tartardiamide, 99%

Specifications

228.25
25g
182° to 186°C
DATD
MFCD00008640
Glass bottle
58477-85-3
C10H16N2O4
≥98.5%
Fine Flakes
White to off-white
100.0
[-CH(OH)CONHCH2CH=CH2]2
98.5
Authentic
99%
N, N'-Diallyl-L-tartardiamide, 99%

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.