missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N'-Diacetylethylenediamine, 98%

Manufacturer:  Alfa Aesar™ A1186618

 View more versions of this product

Catalog No. AAA1186618



Specifications

Specifications

N,N'-Diacetylethylenediamine
172°C to 176°C
50g
MFCD00014966
WNYIBZHOMJZDKN-UHFFFAOYSA-N
N-(2-acetamidoethyl)acetamide
70097
871-78-3
98%
C6H12N2O2
1762229
CC(=O)NCCNC(=O)C
144.174
144.17

Chemical Identifiers

871-78-3
144.174
WNYIBZHOMJZDKN-UHFFFAOYSA-N
N-(2-acetamidoethyl)acetamide
C6H12N2O2
MFCD00014966
70097
CC(=O)NCCNC(=O)C
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents