Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N'-Diacetyl-1,6-diaminohexane, 98+%

$26.16 - $298.62

Chemical Identifiers

Chemical Name or Material N,N'-Diacetyl-1,6-diaminohexane
CAS 3073-59-4
Molecular Formula CH3CONH(CH2)6NHCOCH3
Formula Weight 200.28
MDL Number MFCD00008684
Beilstein 1775764
Synonym N,N'-Diacetyl-1,6-hexanediamine; N,N'-Hexamethylenebis(acetamide)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2277109

 
alfa aesar™
B2277109
10g Each for $26.16

AAB2277118

 
alfa aesar™
B2277118
50g Each for $76.50

AAB2277130

 
alfa aesar™
B2277130
250g Each for $298.62
Description & Specifications

Specifications

Formula Weight 200.28
Chemical Name or Material N,N'-Diacetyl-1,6-diaminohexane
Melting Point 126° to 129°C
Assay ≥98%
Quantity 10g
MDL Number MFCD00008684
CAS 3073-59-4
Beilstein 1775764
Synonym N,N'-Diacetyl-1,6-hexanediamine; N,N'-Hexamethylenebis(acetamide)
RTECS Number AC2976200
Molecular Formula CH3CONH(CH2)6NHCOCH3