missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diacetyl-1,4-phenylenediamine 98.0+%, TCI America™

Supplier:  TCI America D006825G

 View more versions of this product

Catalog No. D006825G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

N,N′-Diacetyl-1,4-phenylenediamine
White-Yellow
MFCD00026142
1,4-diacetamidobenzene, n,n'-diacetyl-1,4-phenylenediamine, n,n'-p-phenylenebisacetamide, p-phenylenediacetamide, n,n'-diacetyl-p-phenylenediamine, 4'-acetamidoacetanilide, 1,4-bisacetamidobenzene, acetamide, n,n'-1,4-phenylenebis, n-4-acetamidophenyl acetamide, n,n'-p-phenylene di acetamide
CC(=O)NC1=CC=C(C=C1)NC(=O)C
192.218
192.22
Crystalline Powder
140-50-1
C10H12N2O2
25 g
KVEDKKLZCJBVNP-UHFFFAOYSA-N
N-(4-acetamidophenyl)acetamide
67324
≥98.0% (GC,N)

Chemical Identifiers

140-50-1
192.218
KVEDKKLZCJBVNP-UHFFFAOYSA-N
67324
CC(=O)NC1=CC=C(C=C1)NC(=O)C
C10H12N2O2
MFCD00026142
1,4-diacetamidobenzene, n,n'-diacetyl-1,4-phenylenediamine, n,n'-p-phenylenebisacetamide, p-phenylenediacetamide, n,n'-diacetyl-p-phenylenediamine, 4'-acetamidoacetanilide, 1,4-bisacetamidobenzene, acetamide, n,n'-1,4-phenylenebis, n-4-acetamidophenyl acetamide, n,n'-p-phenylene di acetamide
N-(4-acetamidophenyl)acetamide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions