Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N'-Di-o-tolylthiourea, 98%

$35.72 - $346.55

Chemical Identifiers

Chemical Name or Material N,N'-Di-o-tolylthiourea
CAS 137-97-3
Molecular Formula C15H16N2S
Formula Weight 256.37
MDL Number MFCD00025922
Beilstein 2125770
Synonym N,N'-Bis(2-Methylphenyl)thiourea
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0363006

  SDS
alfa aesar™
L0363006
5g Each for $35.72

AAL0363022

  SDS
alfa aesar™
L0363022
100g Each for $346.55
Description & Specifications

Specifications

Formula Weight 256.37
Chemical Name or Material N,N'-Di-o-tolylthiourea
MDL Number MFCD00025922
CAS 137-97-3
TSCA TSCA
Melting Point 153° to 155°C
Quantity 5g
Beilstein 2125770
Synonym N,N'-Bis(2-Methylphenyl)thiourea
RTECS Number FE0700000
Assay 98%
Molecular Formula C15H16N2S