Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N'-Di(o-tolyl)ethylenediamine, 98%

$84.97 - $923.57

Chemical Identifiers

Chemical Name or Material N,N'-Di(o-tolyl)ethylenediamine
CAS 94-92-8
Molecular Formula C16H20N2
Formula Weight 240.35
MDL Number MFCD00048073
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2497106

 
alfa aesar™
B2497106
5g Each for $84.97

AAB2497114

 
alfa aesar™
B2497114
25g Each for $286.31

AAB2497122

 
alfa aesar™
B2497122
100g Each for $923.57
Description & Specifications

Specifications

Formula Weight 240.35
Chemical Name or Material N,N'-Di(o-tolyl)ethylenediamine
TSCA TSCA
Melting Point 70° to 73°C
Assay 98%
Quantity 5g
Molecular Formula C16H20N2
MDL Number MFCD00048073
CAS 94-92-8
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.